Specifications
Chemical Name or Material7-Phenylhomopiperazin-5-one
Melting Point104°C to 106°C
CAS89044-79-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more