It is used as a pharmaceutical intermediate of antipsychotics levosulpiride and for the preparation of medicines and other fine chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate of antipsychotics levosulpiride and for the preparation of medicines and other fine chemicals.
Solubility
Soluble in chloroform, methanol. Slightly soluble in water.
Notes
Hygroscopic. Store away from acids, water/moisture, acid chlorides, acid anhydrides and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
RUO – Research Use Only
General References:
- Tomas De Paulis, et al. Potential neuroleptic agents. 4. Chemistry, behavioral pharmacology, and inhibition of [3H]spiperone binding of 3,5-disubstituted N-[(1-ethyl-2-pyrrolidinyl)methyl]-6-methoxysalicylamides.J. Med. Chem.,1986,29(1), 61-69.
- John Meurig Thomas, et al. Bimetallic Catalysts and Their Relevance to the Hydrogen Economy.Ind. Eng. Chem. Res.,2003,42(8), 1563-1570.