Thermo Scientific Chemicals

L-beta-Prolinol, 95%, Thermo Scientific Chemicals

Catalog number: H57199.MD
250 mg, Each
Thermo Scientific Chemicals

L-beta-Prolinol, 95%, Thermo Scientific Chemicals

Catalog number: H57199.MD
250 mg, Each
Quantity
Catalog number: H57199.MD
also known as H57199-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110013-19-9
IUPAC Name
(pyrrolidin-3-yl)methanol
Molecular Formula
C5H11NO
InChI Key
QOTUIIJRVXKSJU-UHFFFAOYNA-N
SMILES
OCC1CCNC1
Form
Fused solid
Assay (GC)
≥94.0%
Proton NMR
Conforms
Appearance (Color)
White to pale yellow.
Optical Rotation
|b = -21? ?3? (C=0.8, CHCl3)

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water

Notes
Hygroscopic. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text