Thermo Scientific Chemicals

Ethyl (S)-(-)-1-Boc-4-oxopiperidine-2-carboxylate, 95%, Thermo Scientific Chemicals

Catalog number: H57213.03
1 g, Each
Thermo Scientific Chemicals

Ethyl (S)-(-)-1-Boc-4-oxopiperidine-2-carboxylate, 95%, Thermo Scientific Chemicals

Catalog number: H57213.03
1 g, Each
Quantity
Catalog number: H57213.03
also known as H57213-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
180854-44-8
IUPAC Name
1-tert-butyl 2-ethyl 4-oxopiperidine-1,2-dicarboxylate
Molecular Formula
C13H21NO5
InChI Key
YAVQLRUBKDCOCI-UHFFFAOYNA-N
SMILES
CCOC(=O)C1CC(=O)CCN1C(=O)OC(C)(C)C
Assay (HPLC)
≥94.0%
Proton NMR
Conforms
Infrared spectrum
Conforms
Appearance (Color)
Pale yellow
Form
Liquid.

Description

1-Boc-4-oxopiperidine-2-carboxylate participates in the one-pot in situ formation and reaction of trimethyl (trichloromethyl) silane which finds application in the synthesis of 2, 2, 2-trichloromethylcarbinols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-4-oxopiperidine-2-carboxylate participates in the one-pot in situ formation and reaction of trimethyl (trichloromethyl) silane which finds application in the synthesis of 2, 2, 2-trichloromethylcarbinols.

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text