Ethyl 2-methylpyridine-4-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals
Ethyl 2-methylpyridine-4-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-methylpyridine-4-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H57239.03
also known as H57239-03
Price (USD)/ Each
98.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
98.60
Add to cart
Ethyl 2-methylpyridine-4-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals
Catalog numberH57239.03
Price (USD)/ Each
98.60
-
Add to cart
Specifications
Chemical Name or MaterialEthyl 2-methylpyridine-4-carboxylate hydrochloride
CAS65550-32-5
Health Hazard 1H318
Health Hazard 2GHS H Statement
H318
Causes serious eye damage.
Health Hazard 3P280-P305+P351+P338-P310
View more
It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
RUO – Research Use Only

General References:

  1. K. B. Prasad.; G. A. Swan. The constitution of yohimbine and related alkaloids. Part X. The synthesis of some 12H-indolo[2 : 3-a]pyridocolinium salts, including flavocoryline and flavopereirine. J. Chem. Soc. 1958, 2024-2038,.
  2. Tathagata Mukherjee.; David G. Hilmey.; Tadhg P. Begley. PLP Catabolism: Identification of the 2-(Acetamidomethylene)succinate Hydrolase Gene in Mesorhizobium loti MAFF303099. Biochemistry. 2008, 47 (23),6233-6241.