It is used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in medicine.
Solubility
Slightly soluble in water
Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials and heat.
RUO – Research Use Only
Kurt Baum.; Phillip T. Berkowitz.; Vytautas Grakauskas.; Thomas G. Archibald. Synthesis of electron-deficient oxetanes. 3-Azidooxetane, 3-nitrooxetane, and 3,3-dinitrooxetane. J. Org. Chem. 1983, 48 (18),2953-2956.
Longwu Ye.; Weimin He.; Liming Zhang. Gold-Catalyzed One-Step Practical Synthesis of Oxetan-3-ones from Readily Available Propargylic Alcohols. J. Am. Chem. Soc. 2010, 132 (25),8550-8551.