DL-beta-Proline hydrochloride, 95%, Thermo Scientific Chemicals
DL-beta-Proline hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-beta-Proline hydrochloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H57365.03
also known as H57365-03
Price (USD)/ Each
89.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
89.00
Add to cart
DL-beta-Proline hydrochloride, 95%, Thermo Scientific Chemicals
Catalog numberH57365.03
Price (USD)/ Each
89.00
-
Add to cart
Specifications
Chemical Name or MaterialDL-beta-Proline hydrochloride
Melting Point30°C to 38°C
CAS953079-94-2
Health Hazard 1H318
Health Hazard 2GHS H Statement
H318
Causes serious eye damage.
View more
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is used as a key intermediate to prepare two different pyrrolidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is used as a key intermediate to prepare two different pyrrolidines.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only
Javier Mendiola, et al. Enzymatic Resolution of N-Substituted-β-prolines.Org. Process Res. Dev.,2009,13(2), 292-296.