3-Bromomethyl-3-oxetanemethanol, 95%, Thermo Scientific Chemicals
3-Bromomethyl-3-oxetanemethanol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromomethyl-3-oxetanemethanol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
250 mg
Catalog number H57413.ME
also known as H57413-ME
Price (USD)/ Each
112.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
132.00 
Save 19.35 (15%)
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
112.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
132.00 
Save 19.35 (15%)
Add to cart
3-Bromomethyl-3-oxetanemethanol, 95%, Thermo Scientific Chemicals
Catalog numberH57413.ME
Price (USD)/ Each
112.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
132.00 
Save 19.35 (15%)
-
Add to cart
Specifications
Chemical Name or Material3-Bromomethyl-3-oxetanemethanol
CAS22633-44-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only