D-Nipecotic acid, 96+%, Thermo Scientific Chemicals
D-Nipecotic acid, 96+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Nipecotic acid, 96+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H57434.06
also known as H57434-06
Price (USD)/ Each
184.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
184.00
Add to cart
D-Nipecotic acid, 96+%, Thermo Scientific Chemicals
Catalog numberH57434.06
Price (USD)/ Each
184.00
-
Add to cart
Chemical Identifiers
CAS25137-00-2
IUPAC Namepiperidine-3-carboxylic acid
Molecular FormulaC6H11NO2
InChI KeyXJLSEXAGTJCILF-UHFFFAOYNA-N
SMILESOC(=O)C1CCCNC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay from Supplier's CofA≥96.0%
FormPowder
Melting Point251-255?C
Assay from Supplier's CofAH₂O ≤ 0.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only