Thermo Scientific Chemicals

1-Oxa-6-azaspiro[3.4]octane oxalate, 96%, Thermo Scientific Chemicals

Catalog number: H57452.ME
500 mg, Each
Thermo Scientific Chemicals

1-Oxa-6-azaspiro[3.4]octane oxalate, 96%, Thermo Scientific Chemicals

Catalog number: H57452.ME
500 mg, Each
Quantity
Catalog number: H57452.ME
also known as H57452-ME
Price (USD)
Quantity
-
Have Questions?
Form
Fused solid
Appearance (Color)
White to cream
Assay (GC)
≥95.0%
Proton NMR
Conforms

Description

1-Oxa-6-azaspiro[3.4]octane oxalate is used as a pharmaceutical intermediate in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Oxa-6-azaspiro[3.4]octane oxalate is used as a pharmaceutical intermediate in synthetic chemistry.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text