1-Boc-3-(bromomethyl)azetidine, 95%, Thermo Scientific Chemicals
1-Boc-3-(bromomethyl)azetidine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Boc-3-(bromomethyl)azetidine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
500 mg
Catalog number H57492.MD
also known as H57492-MD
Price (USD)/ Each
111.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
111.00
Add to cart
1-Boc-3-(bromomethyl)azetidine, 95%, Thermo Scientific Chemicals
Catalog numberH57492.MD
Price (USD)/ Each
111.00
-
Add to cart
Specifications
Chemical Name or Material1-Boc-3-(bromomethyl)azetidine
CAS253176-93-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Sparingly soluble in water at 25°C, 0.83 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only