Thermo Scientific Chemicals

Oxetane-3-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H57578.MD
250 mg, Each
Thermo Scientific Chemicals

Oxetane-3-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H57578.MD
250 mg, Each
Quantity
Catalog number: H57578.MD
also known as H57578-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
114012-41-8
IUPAC Name
oxetane-3-carboxylic acid
Molecular Formula
C4H6O3
InChI Key
UWOTZNQZPLAURK-UHFFFAOYSA-N
SMILES
OC(=O)C1COC1
Appearance (Color)
Pale yellow
Form
Liquid
Proton NMR
Conforms to structure
Purity
≥94.0%

Description

Oxetane-3-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxetane-3-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Moisture sensitive. . Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text