Thermo Scientific Chemicals

2-Oxa-6-azaspiro[3.4]octane, 95%, Thermo Scientific Chemicals

Catalog number: H57579.MD
250 mg, Each
Thermo Scientific Chemicals

2-Oxa-6-azaspiro[3.4]octane, 95%, Thermo Scientific Chemicals

Catalog number: H57579.MD
250 mg, Each
Quantity
Catalog number: H57579.MD
also known as H57579-MD
Price (USD)
Quantity
-
Have Questions?

Description

2-Oxa-6-azaspiro[3.4]octane is involved in the preparation of azaspirocycle or azetidine substituted 4-anilinoquinazoline derivatives, which exhibits epidermal growth factor receptor (EGFR) inhibitory activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Oxa-6-azaspiro[3.4]octane is involved in the preparation of azaspirocycle or azetidine substituted 4-anilinoquinazoline derivatives, which exhibits epidermal growth factor receptor (EGFR) inhibitory activities.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids, reducing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text