Thermo Scientific Chemicals

(S)-(-)-1-Boc-4-oxopiperidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H57617.03
1 g, Each
Thermo Scientific Chemicals

(S)-(-)-1-Boc-4-oxopiperidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H57617.03
1 g, Each
Quantity
Catalog number: H57617.03
also known as H57617-03
Price (USD)
Quantity
-
Have Questions?

Description

It is a fine chemical and synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a fine chemical and synthetic intermediate.

Solubility
Soluble (12 g/L) (25°C)

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text