Thermo Scientific Chemicals
3-Methoxyazetidine hydrochloride, 95%, Thermo Scientific Chemicals
Catalog number: H57769.ME
500 mg, Each
Thermo Scientific Chemicals
3-Methoxyazetidine hydrochloride, 95%, Thermo Scientific Chemicals
Catalog number: H57769.ME
500 mg, Each
Quantity
Catalog number: H57769.ME
also known as H57769-ME
Price (USD)
Quantity
-
Have Questions?
Description
It is used in the preparation of isoxazole-thiazole derivatives as GABA. It is a receptor inverse agonists for use in the treatment of cognitive disorders.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the preparation of isoxazole-thiazole derivatives as GABA. It is a receptor inverse agonists for use in the treatment of cognitive disorders.
Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials and heat.