N-Boc-L-beta-proline is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-L-beta-proline is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water (slightly).
Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only
Rajesh B. M.; Madhushree Bhattacharya; Biswadip Banerji and Javed Iqbal. Synthesis of L-proline derived cyclic β-turn mimics via ring closing metathesis. ARKIVOC. 2004, 111-120.