2-n-Propyl-1-pentanol, 97%, Thermo Scientific Chemicals
2-n-Propyl-1-pentanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-n-Propyl-1-pentanol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H57954.06
also known as H57954-06
Price (USD)/ Each
145.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
145.00
Add to cart
2-n-Propyl-1-pentanol, 97%, Thermo Scientific Chemicals
Catalog numberH57954.06
Price (USD)/ Each
145.00
-
Add to cart
Chemical Identifiers
CAS58175-57-8
IUPAC Name2-propylpentan-1-ol
Molecular FormulaC8H18O
InChI KeyLASHFHLFDRTERB-UHFFFAOYSA-N
SMILESCCCC(CO)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Purity≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only