This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Cyanophenylzinc bromide is used to produce biphenyl-4-carbonitrile.
Solubility
Reacts with water.
Notes
Incompatible with oxidizing agents, air, bases and heat. Air Sensitive. Store under dry iner
RUO – Research Use Only
Anne Eeg Jensen; Florian Kneisel; Paul Knochel. Ethyl 3-(p-Cyanophenyl)Propionate from Ethyl 3-Iodopropionate and p-Cyanophenylzinc Bromide. Organic Syntheses.2003, 35