This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Thienylzinc bromide is used as medical intermediates.
Solubility
Reacts with water.
Notes
Air Sensitive. Store at 4°C. Incompatible with oxidizing agents, acid chlorides. Store under dry inert
RUO – Research Use Only
Jie Wu and Zhen Yang. Nickel-Catalyzed Cross-Couplings of 4-Diethylphosphonooxycoumarins with Organozinc Reagents: An Efficient New Methodology for the Synthesis of 4-Substituted Coumarins. J. Org. Chem.2001, 66 (23), 7875-7878.