Thermo Scientific Chemicals

1-Adamantylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: H58408.AD
50 mL, Each
Thermo Scientific Chemicals

1-Adamantylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: H58408.AD
50 mL, Each
Quantity
Catalog number: H58408.AD
also known as H58408-AD
Price (USD)
Quantity
-
Have Questions?

Description

1-Adamantylzinc bromide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantylzinc bromide is used as pharmaceutical intermediates.

Solubility
Reacts with water.

Notes
Air Sensitive. Incompatible with oxidizing agents, bases and heat. Store under dry inert gas. Sto
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text