Thermo Scientific Chemicals

6-Chlorobenzimidazole-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H58648.03
1 g, Each
Thermo Scientific Chemicals

6-Chlorobenzimidazole-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H58648.03
1 g, Each
Quantity
Catalog number: H58648.03
also known as H58648-03
Price (USD)
Quantity
-
Have Questions?

Description

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
This products exists as two tautomers as there is a rapid exchange of the proton between the two heterocyclic nitrogens. This renders the 5- and 6- positions on the ring chemically equivalent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text