It is used in the cross-coupling reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the cross-coupling reaction.
Solubility
Reacts with water.
Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids and wate
RUO – Research Use Only
Tamio Hayashi.; Akihiro Yamamoto.; Toshiya Hagihara. Stereo- and regiochemistry in palladium-catalyzed nucleophilic substitution of optically active (E)- and (Z)-allyl acetates. J. Org. Chem. 1986, 51 (5),723-727.
Mario Rottländer.; Paul Knochel. Palladium-Catalyzed Cross-Coupling Reactions with Aryl Nonaflates: A Practical Alternative to Aryl Triflates. J. Org. Chem. 1998, 63 (1),203-208.