4-Bromobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
4-Bromobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 mL
Catalog number H58662.AD
also known as H58662-AD
Price (USD)/ Each
226.00
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
226.00
Add to cart
4-Bromobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog numberH58662.AD
Price (USD)/ Each
226.00
-
Add to cart
Chemical Identifiers
CAS115055-85-1
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Concentration0.40-0.60M
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only