Thermo Scientific Chemicals

(1,3-Dioxolan-2-ylmethyl)zinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: H58702.AD
50 mL, Each
Thermo Scientific Chemicals

(1,3-Dioxolan-2-ylmethyl)zinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: H58702.AD
50 mL, Each
Quantity
Catalog number: H58702.AD
also known as H58702-AD
Price (USD)
Quantity
-
Have Questions?
Form
Liquid
Concentration
0.4-0.6M

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Reacts with water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text