Specifications
Chemical Name or Materialsec-Butylzinc bromide
Name NotePackaged under Argon in resealable ChemSeal™ bottles
CAS171860-66-5
Health Hazard 1H225-H260-H302-H314-H335-H336-H351
Health Hazard 2GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
View more