Cyclopentylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Cyclopentylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopentylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 mL
Catalog number H58764.AD
also known as H58764-AD
Price (USD)/ Each
249.00
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
249.00
Add to cart
Cyclopentylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog numberH58764.AD
Price (USD)/ Each
249.00
-
Add to cart
Specifications
Chemical Name or MaterialCyclopentylzinc bromide
Name NotePackaged under Argon in resealable ChemSeal™ bottles
CAS171860-68-7
Health Hazard 1H225-H260-H302-H314-H335-H336-H351
Health Hazard 2GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an active nucleophile in Pd-catalyzed cross-coupling of heteroaryl halides.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Keith Biggadike.; Mohamed Boudjelal.; Margaret Clackers.; Diane M. Coe.; Derek A. Demaine.; George W. Hardy.; Davina Humphreys.; Graham G. A. Inglis.; Michael J. Johnston.; Haydn T. Jones.; David House.; Richard Loiseau.; Deborah Needham.; Philip A. Skone, Iain Uings.; Gemma Veitch.; Gordon G. Weingarten.; Iain M. McLay.; Simon J. F. Macdonald. Nonsteroidal Glucocorticoid Agonists: Tetrahydronaphthalenes with Alternative Steroidal A-Ring Mimetics Possessing Dissociated (Transrepression/Transactivation) Efficacy Selectivity. J. Med. Chem. 2007, 50 (26),6519-6534.