3-Methylamino-1-propanol is an alkanolamine that is used to redisperse compacted solids (such as antiperspirant materials) in order to analyze its components.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methylamino-1-propanol is an alkanolamine that is used to redisperse compacted solids (such as antiperspirant materials) in order to analyze its components.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Yuki Inoue; Atsushi Kuwahara; Kohei Ohmori; Hirobumi Sunayama; Tooru Ooya; Toshifumi Takeuchi. Fluorescent molecularly imprinted polymer thin films for specific protein detection prepared with dansyl ethylenediamine-conjugated O-acryloyl l-hydroxyproline. Biosensors and Bioelectronics,2013, 48113-119.
- T. Lindgren; R. Sillanpää; T. Nortia; K. Pihlaja. Copper(II) complexes with some N-methylated 3-amino-1-propanols. Inorganica Chimica Acta,1984, 82(1), 1-4.