Cyclohexylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Cyclohexylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclohexylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 mL
Catalog number H58852.AD
also known as H58852-AD
Price (USD)/ Each
195.65
Online exclusive
217.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
195.65
Online exclusive
217.00 
Save 21.35 (10%)
Add to cart
Cyclohexylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog numberH58852.AD
Price (USD)/ Each
195.65
Online exclusive
217.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7565-57-3
IUPAC Namebromozincylium; cyclohexanide
Molecular FormulaC6H11BrZn
InChI KeyPVURAUIMVICLOH-UHFFFAOYSA-M
SMILES[Zn+]Br.C1CC[CH-]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Concentration0.4-0.6M
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as primary and secondary intermediate.

Solubility
Reacts with water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Maria F. Dalence-Guzmán.; Jörgen Toftered.; Viveca Thornqvist Oltner.; David Wensbo.; Martin H. Johansson. Synthesis of novel tetrahydroisoquinoline bronchodilators.Bioorg. Med. Chem. Lett. 2010, 20 (17),4999-5003.
  2. Jie Wu , Yun Liao.; Zhen Yang. Synthesis of 4-Substituted Coumarins via the Palladium-Catalyzed Cross-Couplings of 4-Tosylcoumarins with Terminal Acetylenes and Organozinc Reagents.J. Org. Chem. 2001, 66 (10),3642-3645.