Thermo Scientific Chemicals

1,6-Bismaleimidohexane, 97%, Thermo Scientific Chemicals

Catalog number: H58874.ME
500 mg, Each
Thermo Scientific Chemicals

1,6-Bismaleimidohexane, 97%, Thermo Scientific Chemicals

Catalog number: H58874.ME
500 mg, Each
Quantity
Catalog number: H58874.ME
also known as H58874-ME
Price (USD)
Quantity
-
Have Questions?

Description

A sulfhydryl reactive homobifunctional cross linking reagent. Under mild conditions, will permit irreversible cross-linking of sulfhydral containing compounds at a pH range of 6.5 to 7.5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A sulfhydryl reactive homobifunctional cross linking reagent. Under mild conditions, will permit irreversible cross-linking of sulfhydral containing compounds at a pH range of 6.5 to 7.5.

Solubility
Soluble in chloroform.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text