Specifications
Chemical Name or Material4-Cyanobutylzinc bromide
Name NotePackaged under Argon in resealable ChemSeal™ bottles
CAS226570-68-9
Health Hazard 1H225-H302-H314-H335-H336-H351
Health Hazard 2GHS H Statement
H225-H261-H314-H351-H302-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
View more