This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications It is employed as a intermediate for pharmaceutical.
Solubility Reacts with water.
Notes Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-vent