Thermo Scientific Chemicals

N-Fmoc-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H59007.06
5 g, Each
Thermo Scientific Chemicals

N-Fmoc-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H59007.06
5 g, Each
Quantity
Catalog number: H59007.06
also known as H59007-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71989-14-5
IUPAC Name
(2S)-4-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoate
Molecular Formula
C23H24NO6
InChI Key
FODJWPHPWBKDON-IBGZPJMESA-M
SMILES
CC(C)(C)OC(=O)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O
Appearance (Color)
White to pale cream
Form
Powder
Assay (HPLC)
>97.5%
Optical Rotation
-24? ±2? (c=1 in DMF)

Description

L-Aspartic acid is a nonessential amino acid that is used to biosynthesize other amino acids within the human body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid is a nonessential amino acid that is used to biosynthesize other amino acids within the human body.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text