Chemical Identifiers
CAS71989-18-9
IUPAC Name5-(tert-butoxy)-4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoic acid
Molecular FormulaC24H27NO6
InChI KeyGOPWHXPXSPIIQZ-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more