1-n-Butyl-3-methylimidazolium dicyanamide, 97%, Thermo Scientific Chemicals
1-n-Butyl-3-methylimidazolium dicyanamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-n-Butyl-3-methylimidazolium dicyanamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Catalog number H59175.18
also known as H59175-18
Price (USD)/ Each
806.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
806.00
Add to cart
1-n-Butyl-3-methylimidazolium dicyanamide, 97%, Thermo Scientific Chemicals
Catalog numberH59175.18
Price (USD)/ Each
806.00
-
Add to cart
Chemical Identifiers
CAS448245-52-1
IUPAC Name1-butyl-3-methyl-1H-imidazol-3-ium; [(cyanoimino)methylidene]azanide
Molecular FormulaC10H15N5
InChI KeyICIVTHOGIQHZRY-UHFFFAOYSA-N
SMILES[N-]=C=NC#N.CCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only