Thermo Scientific Chemicals

Ethyl 3-ethoxyacrylate, 98%, Thermo Scientific Chemicals

Catalog number: H59187.09
10 g, Each
Thermo Scientific Chemicals

Ethyl 3-ethoxyacrylate, 98%, Thermo Scientific Chemicals

Catalog number: H59187.09
10 g, Each
Quantity
Catalog number: H59187.09
also known as H59187-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1001-26-9
IUPAC Name
ethyl (2E)-3-ethoxyprop-2-enoate
Molecular Formula
C7H12O3
InChI Key
ITQFPVUDTFABDH-AATRIKPKSA-N
SMILES
CCOC=CC(=O)OCC
Assay (HPLC)
>97.5%

Description

Used in the preparation of N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-5-yl)phenyl]-N-ethylacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-5-yl)phenyl]-N-ethylacetamide.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text