Thermo Scientific Chemicals

1-Ethyl-2,3-dimethylimidazolium chloride, 97%, Thermo Scientific Chemicals

Catalog number: H59225.18
50 g, Each
Thermo Scientific Chemicals

1-Ethyl-2,3-dimethylimidazolium chloride, 97%, Thermo Scientific Chemicals

Catalog number: H59225.18
50 g, Each
Quantity
Catalog number: H59225.18
also known as H59225-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92507-97-6
IUPAC Name
1-ethyl-2,3-dimethyl-1H-imidazol-3-ium
Molecular Formula
C7H13N2
InChI Key
IRGDPGYNHSIIJJ-UHFFFAOYSA-N
SMILES
CCN1C=C[N+](C)=C1C
Proton NMR
>96.0%

Description

It is used reaction media for chemical processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used reaction media for chemical processes.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text