It is used as a pharmaceutical, chemical, paint and dye intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical, chemical, paint and dye intermediate.
Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only
Ishiwata Hiroyuki.; Sone Hiroki.; Kigoshi Hideo.; Yamada Kiyoyuki. Enantioselective total synthesis of doliculide, a potent cytotoxic cyclodepsipeptide of marine origin and structure-cytotoxicity relationships of synthetic doliculide congeners. Tetrahedron. 1994, 50 (45),12853-12882.
Andrew G. Myers.; James L. Gleason.; Taeyoung Yoon. Practical method for the synthesis of D- or L-.alpha.-amino acids by the alkylation of (+)- or (-)-pseudoephedrine glycinamide. J. Am. Chem. Soc. 1995, 117 (32),8488-8489.