It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Partly soluble in water and chloroform.
Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Jens U. Nielsen.; Maria J. Esplandiu.; Dieter M. Kolb. 4-Nitrothiophenol SAM on Au(111) Investigated by in Situ STM, Electrochemistry, and XPS. Langmuir. 2001, 17 (11), 3454-3459.
- Hiromori Tsutsumi.; Shozo Furumoto.; Masayuki Morita.; Yoshiharu Matsuda. Electrochemical Behavior of a 4-Nitrothiophenol Modified Electrode Prepared by the Self-Assembly Method. J. Colloid Interface Sc. 1995, 171 (2),505-511.