Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium acetate, 97%, Thermo Scientific Chemicals

Catalog number: H59277.06
5 g, Each
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium acetate, 97%, Thermo Scientific Chemicals

Catalog number: H59277.06
5 g, Each
Quantity
Catalog number: H59277.06
also known as H59277-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143314-17-4
IUPAC Name
1-ethyl-3-methyl-1H-imidazol-3-ium acetate
Molecular Formula
C8H14N2O2
InChI Key
XIYUIMLQTKODPS-UHFFFAOYSA-M
SMILES
CC([O-])=O.CCN1C=C[N+](C)=C1
Assay from Supplier's CofA
≥96.0% (HPLC)
Appearance (Color)
Pale yellow to orange to brown
Assay from Supplier's CofA
≥96.0% (HPLC)
Form
Liquid
Appearance (Color)
Pale yellow to orange to brown

Description

It is used as a solvent and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a solvent and as pharmaceutical intermediate.

Solubility
Soluble in water, acetonitrile, acetone, dichloromethane.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text