3-Aminoazetidine dihydrochloride, 95%, Thermo Scientific Chemicals
3-Aminoazetidine dihydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminoazetidine dihydrochloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
250 mg
Catalog number H59318.ME
also known as H59318-ME
Price (USD)/ Each
279.00
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
279.00
Add to cart
3-Aminoazetidine dihydrochloride, 95%, Thermo Scientific Chemicals
Catalog numberH59318.ME
Price (USD)/ Each
279.00
-
Add to cart
Specifications
Chemical Name or Material3-Aminoazetidine dihydrochloride
CAS102065-89-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
3-Aminoazetidine dihydrochloride, is used as active pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminoazetidine dihydrochloride, is used as active pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only