Thermo Scientific Chemicals

4-Fluoro-1-indanone, 97%, Thermo Scientific Chemicals

Catalog number: H59356.06
5 g, Each
Thermo Scientific Chemicals

4-Fluoro-1-indanone, 97%, Thermo Scientific Chemicals

Catalog number: H59356.06
5 g, Each
Quantity
Catalog number: H59356.06
also known as H59356-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
699-99-0
IUPAC Name
4-fluoro-2,3-dihydro-1H-inden-1-one
Molecular Formula
C9H7FO
InChI Key
HOMSJDBZHCPYHY-UHFFFAOYSA-N
SMILES
FC1=CC=CC2=C1CCC2=O
Assay (HPLC)
>96.0%

Description

It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. 

Solubility
Soluble in alcohol and chloroform

Notes
Store at room temperature. Keep away from oxidizing agents and heats.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text