Thermo Scientific Chemicals

Methyl pyrrole-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H59437.06
5 g, Each
Thermo Scientific Chemicals

Methyl pyrrole-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H59437.06
5 g, Each
Quantity
Catalog number: H59437.06
also known as H59437-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1193-62-0
IUPAC Name
methyl 1H-pyrrole-2-carboxylate
Molecular Formula
C6H7NO2
InChI Key
VONGYFFEWFJHNP-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CN1
Assay (HPLC)
>96.0%

Description

It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. 

Solubility
Soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text