Ethyl glutaryl chloride, 97%, Thermo Scientific Chemicals
Ethyl glutaryl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl glutaryl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Catalog number H59443.AA
also known as H59443-AA
Price (USD)/ Each
54.90
-
Add to cart
Quantity:
5 mL
Request bulk or custom format
Price (USD)/ Each
54.90
Add to cart
Ethyl glutaryl chloride, 97%, Thermo Scientific Chemicals
Catalog numberH59443.AA
Price (USD)/ Each
54.90
-
Add to cart
Chemical Identifiers
CAS5205-39-0
IUPAC Nameethyl 5-chloro-5-oxopentanoate
Molecular FormulaC7H11ClO3
InChI KeyKKJAQUGGQMCNJY-UHFFFAOYSA-N
SMILESCCOC(=O)CCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR>96.0%
Ethyl glutaryl chloride is a main cationic monomer, it can bring quaternary ammonium salt

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl glutaryl chloride is a main cationic monomer, it can bring quaternary ammonium salt

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. R. Breslow.; A. W. Czarnik.; M. Lauer.; R. Leppkes.; J. Winkler.; S. Zimmerman. Mimics of transaminase enzymes. J. Am. Chem. Soc. 1986, 108, (8), 1969-1979.
  2. David R. Carbery.; Serge Reignier.; Neil D. Miller.; Harry Adams.; Joseph P. A. Harrity. Investigation of a Stereoselective Co-Mediated Rearrangement Reaction. J. Org. Chem. 2003, 68, (11), 4392-4399.