Thermo Scientific Chemicals

Ethyl glutaryl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H59443.AC
25 mL, Each
Thermo Scientific Chemicals

Ethyl glutaryl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H59443.AC
25 mL, Each
Quantity
Catalog number: H59443.AC
also known as H59443-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5205-39-0
IUPAC Name
ethyl 5-chloro-5-oxopentanoate
Molecular Formula
C7H11ClO3
InChI Key
KKJAQUGGQMCNJY-UHFFFAOYSA-N
SMILES
CCOC(=O)CCCC(Cl)=O
Proton NMR
>96.0%

Description

Ethyl glutaryl chloride is a main cationic monomer, it can bring quaternary ammonium salt

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl glutaryl chloride is a main cationic monomer, it can bring quaternary ammonium salt

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text