Thermo Scientific Chemicals

N-Thionylaniline, 96%, Thermo Scientific Chemicals

Catalog number: H59451.14
25 g, Each
Thermo Scientific Chemicals

N-Thionylaniline, 96%, Thermo Scientific Chemicals

Catalog number: H59451.14
25 g, Each
Quantity
Catalog number: H59451.14
also known as H59451-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1122-83-4
IUPAC Name
N-(oxo-λ⁴-sulfanylidene)aniline
Molecular Formula
C6H5NOS
InChI Key
FIOJWGRGPONADF-UHFFFAOYSA-N
SMILES
O=S=NC1=CC=CC=C1
Assay (GC)
≥95.0%

Description

It is a versatile synthetic intermediate and an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a versatile synthetic intermediate and an active pharmaceutical intermediate.

Solubility
Partly miscible in water.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text