Thermo Scientific Chemicals

Ethyl 2-oxocyclohexanecarboxylate, 95%, Thermo Scientific Chemicals

Catalog number: H59495.14
25 g, Each
Thermo Scientific Chemicals

Ethyl 2-oxocyclohexanecarboxylate, 95%, Thermo Scientific Chemicals

Catalog number: H59495.14
25 g, Each
Quantity
Catalog number: H59495.14
also known as H59495-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1655-07-8
IUPAC Name
ethyl 2-oxocyclohexane-1-carboxylate
Molecular Formula
C9H14O3
InChI Key
FGSGHBPKHFDJOP-UHFFFAOYNA-N
SMILES
CCOC(=O)C1CCCCC1=O
Refractive Index
1.4775-1.4800 @ 20?C
Comment
Product exists as a mixture of keto-enol tautomers due to formation of a stable quaternary substituted double bond.
Appearance (Color)
Clear colorless to pale yellow
Assay (unspecified)
≥94.0% (sum of tautomers)
Form
Liquid

Description

Ethyl 2-oxocyclohexanecarboxylate is used to produce other chemicals. For example, it can react with 4-methyl-aniline to get 2-oxo-cyclohexanecarboxylic acid p-toluidide. The reaction occurs with reagent DMAP

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-oxocyclohexanecarboxylate is used to produce other chemicals. For example, it can react with 4-methyl-aniline to get 2-oxo-cyclohexanecarboxylic acid p-toluidide. The reaction occurs with reagent DMAP

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text