Thermo Scientific Chemicals

1-Methyl-3-n-octylimidazolium chloride, 97%, Thermo Scientific Chemicals

Catalog number: H59534.06
5 g, Each
Thermo Scientific Chemicals

1-Methyl-3-n-octylimidazolium chloride, 97%, Thermo Scientific Chemicals

Catalog number: H59534.06
5 g, Each
Quantity
Catalog number: H59534.06
also known as H59534-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64697-40-1
IUPAC Name
3-methyl-1-octyl-1H-imidazol-3-ium chloride
Molecular Formula
C12H23ClN2
InChI Key
OXFBEEDAZHXDHB-UHFFFAOYSA-M
SMILES
[Cl-].CCCCCCCCN1C=C[N+](C)=C1
Proton NMR
>96.0%

Description

Role of 1-methyl-3-octylimidazolium chloride in the micellization behavior of amphiphilic drug amitriptyline hydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Role of 1-methyl-3-octylimidazolium chloride in the micellization behavior of amphiphilic drug amitriptyline hydrochloride.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text