It is an intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an intermediate in organic syntheses.
Solubility
Soluble in water. (5g/100ml)
Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Gianfranco Balbonia.; Mauro Marastoni.; Stefania Merighi.; Pier Andrea Borea.; Roberto Tomatis. Synthesis and activity of 3-pyridylamine ligands at central nicotinic receptors. Eur. J. Med. Chem. 2000, 35 (11),979-988.
Hardwin O’Dowd.; Jason G. Lewis.; Joaquim Trias.; Rumi Asano.; Johanne Blais.; Sara L. Lopez.; Craig K. Park.; Charlotte Wu.; Wen Wang.; Mikhail F. Gordeev. Novel antibacterial azetidine lincosamides .Bioorg. Med. Chem. Lett. 2008, 18 (8),2645-2648.