3-(Boc-amino)azetidine, 98%, Thermo Scientific Chemicals
3-(Boc-amino)azetidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(Boc-amino)azetidine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
250 mg
Catalog number H59556.ME
also known as H59556-ME
Price (USD)/ Each
146.00
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
146.00
Add to cart
3-(Boc-amino)azetidine, 98%, Thermo Scientific Chemicals
Catalog numberH59556.ME
Price (USD)/ Each
146.00
-
Add to cart
Specifications
Chemical Name or Material3-(Boc-amino)azetidine
CAS91188-13-5
Boiling Point268°C
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
It is an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in organic syntheses.

Solubility
Soluble in water. (5g/100ml)

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Gianfranco Balbonia.; Mauro Marastoni.; Stefania Merighi.; Pier Andrea Borea.; Roberto Tomatis. Synthesis and activity of 3-pyridylamine ligands at central nicotinic receptors. Eur. J. Med. Chem. 2000, 35 (11),979-988.
  2. Hardwin O’Dowd.; Jason G. Lewis.; Joaquim Trias.; Rumi Asano.; Johanne Blais.; Sara L. Lopez.; Craig K. Park.; Charlotte Wu.; Wen Wang.; Mikhail F. Gordeev. Novel antibacterial azetidine lincosamides .Bioorg. Med. Chem. Lett. 2008, 18 (8),2645-2648.