4-Methoxy-N-phenylbenzamide, 97%, Thermo Scientific Chemicals
4-Methoxy-N-phenylbenzamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxy-N-phenylbenzamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H59690.MD
also known as H59690-MD
Price (USD)/ Each
152.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
152.00
Add to cart
4-Methoxy-N-phenylbenzamide, 97%, Thermo Scientific Chemicals
Catalog numberH59690.MD
Price (USD)/ Each
152.00
-
Add to cart
Chemical Identifiers
CAS7465-88-5
IUPAC Name4-methoxy-N-phenylbenzamide
Molecular FormulaC14H13NO2
InChI KeyXOJAJRFBOKCXPI-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(=O)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only